missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Septin-11 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32159FR
This item is not returnable.
View return policy
Description
Septin-11 Polyclonal antibody specifically detects Septin-11 in Human, Mouse samples. It is validated for Western Blot
Specifications
| Septin-11 | |
| Polyclonal | |
| Western Blot | |
| FLJ10849, septin 11, septin-11 | |
| The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Septin 11 using the numbering given in entry NP_060713.1 (GeneID 55752). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 55752 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction