missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Septin-10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583886
This item is not returnable.
View return policy
Description
Immunogen sequence: MASSEVARHL LFQSHMATKT TCMSSQGSDD EQIKRENIRS LTMSGHVG Highest antigen sequence indentity to the following orthologs: Rat - 31%, Mouse - 31%.
RAPGEF6 is a guanine nucleotide exchange factor (GEF) for Rap1A, Rap2A and M-Ras GTPases. RAPGEF6 does not interact with cAMP.
Specifications
| Septin-10 | |
| Polyclonal | |
| Unconjugated | |
| SEPT10 | |
| 4921515A04Rik; 9430099J10Rik; AA408298; AI874685; FLJ11619; SEPT10; septin 10; SEPTIN10; septin-10 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 151011 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9P0V9 | |
| SEPT10 | |
| Recombinant protein corresponding to Human SEPT10. Recombinant protein control fragment (Product #RP-100558). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction