missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Septin-10 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-44288FR
This item is not returnable.
View return policy
Description
Septin-10 Polyclonal antibody specifically detects Septin-10 in Human samples. It is validated for Western Blot
Specifications
| Septin-10 | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Immunogen affinity purified | |
| 151011 | |
| Purified |
| Polyclonal | |
| Western Blot | |
| Rabbit | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction