missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Septin-10 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-44287CL1
This item is not returnable.
View return policy
Description
Septin-10 Polyclonal antibody specifically detects Septin-10 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| Septin-10 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| FLJ11619, septin 10, septin-10 | |
| The epitope recognized by this antibody maps to a region between residue 404 to 454 of human Septin-10 using the numbering given in entry Q9P0V9.2 (GeneID 151011). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 151011 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction