Learn More
Septin-10 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-44287AF750
This item is not returnable.
View return policy
Description
Septin-10 Polyclonal antibody specifically detects Septin-10 in Human samples. It is validated for Western Blot
Specifications
| Septin-10 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Cell Cycle and Replication | |
| 151011 |
| Polyclonal | |
| FLJ11619, septin 10, septin-10 | |
| The epitope recognized by this antibody maps to a region between residue 404 to 454 of human Septin-10 using the numbering given in entry Q9P0V9.2 (GeneID 151011). | |
| Primary | |
| Human |