Learn More
Septin-10 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-44288AF594
This item is not returnable.
View return policy
Description
Septin-10 Polyclonal antibody specifically detects Septin-10 in Human samples. It is validated for Western Blot
Specifications
| Septin-10 | |
| Polyclonal | |
| Western Blot | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 594 | |
| RUO | |
| Purified |