missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Septin 1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599715
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Septin 1.
Septin 1 (SEPT1) is a member of the septin family of GTPases. Members of this family are required for cytokinesis and the maintenance of cellular morphology. This gene encodes a protein that can form homo- and heterooligomeric filaments, and may contribute to the formation of neurofibrillary tangles in Alzheimer's disease. Alternatively spliced transcript variants have been found but the full-length nature of these variants has not been determined.
Specifications
| Septin 1 | |
| Polyclonal | |
| Unconjugated | |
| SEPTIN1 | |
| 1-Sep; DIFF6; differentiation 6; differentiation 6 (deoxyguanosine triphosphate triphosphohydrolase); differentiation protein 6; LARP; MGC20394; Peanut-like protein 3; PNUTL3; Protein Diff6; SEP1; Sept1; septin 1; Septin1; septin-1; Serologically defined breast cancer antigen NY-BR-24 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1731 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8WYJ6 | |
| SEPTIN1 | |
| A synthesized peptide derived from human SEPT1(Accession Q8WYJ6), corresponding to amino acid residues E279-T329. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction