missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Septin-1 Antibody (OTI1E1), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74072CL1
This item is not returnable.
View return policy
Description
Septin-1 Monoclonal antibody specifically detects Septin-1 in Human samples. It is validated for Western Blot
Specifications
| Septin-1 | |
| Monoclonal | |
| CoraFluor 1 | |
| DIFF6SEP1, differentiation 6 (deoxyguanosine triphosphate triphosphohydrolase), LARP, Peanut-like protein 3, PNUTL3MGC20394, septin 1, septin-1, Serologically defined breast cancer antigen NY-BR-24 | |
| Full length human recombinant protein of human SEPT1(NP_443070) produced in HEK293T cell. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 1731 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
| Western Blot | |
| OTI1E1 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction