missing translation for 'onlineSavingsMsg'
Learn More

Septin-1 Antibody, DyLight 680, Novus Biologicals™

Product Code. 18422676 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18422676 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18422676 Supplier Novus Biologicals Supplier No. NBP281892FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Septin-1 Polyclonal antibody specifically detects Septin-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Septin-1
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias DIFF6SEP1, differentiation 6 (deoxyguanosine triphosphate triphosphohydrolase), LARP, Peanut-like protein 3, PNUTL3MGC20394, septin 1, septin-1, Serologically defined breast cancer antigen NY-BR-24
Host Species Rabbit
Immunogen Septin-1 antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human Septin-1. The immunogen is located within amino acids 300 - 350 of Septin-1. Amino Acid Squence: SRQSATEIPLPMLPLAD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 1731
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.