missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Septin-1 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81892AF488
This item is not returnable.
View return policy
Description
Septin-1 Polyclonal antibody specifically detects Septin-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| Septin-1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 488 | |
| DIFF6SEP1, differentiation 6 (deoxyguanosine triphosphate triphosphohydrolase), LARP, Peanut-like protein 3, PNUTL3MGC20394, septin 1, septin-1, Serologically defined breast cancer antigen NY-BR-24 | |
| Septin-1 antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human Septin-1. The immunogen is located within amino acids 300 - 350 of Septin-1. Amino Acid Squence: SRQSATEIPLPMLPLAD | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 1731 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction