missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Septa for 384-Well Plates, for 3500/SeqStudio™ Flex

Product Code. 15819856 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
20 each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15819856 20 each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15819856 Supplier Applied Biosystems™ Supplier No. 4412520
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

These septa for 384-well plates are for use with Applied Biosystems 3500 and SeqStudio Flex Series Genetic Analyzers.

These septa for 384-well plates are for use with Applied Biosystems 3500 and SeqStudio Flex series genetic analyzers.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 20 each
Format 384-well Plate
For Use With (Equipment) 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers
Product Type Septa Plate
Shipping Condition Room Temperature
Content And Storage This product contains 20 septa.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.