missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Septa for 0.5 mL sample tubes (for the 310 Genetic Analyzer)

Product Code. 15819726 Shop All Applied Biosystems Products
Click to view available options
Quantity:
500 Each
Unit Size:
Pack of 500
This item is not returnable. View return policy

Product Code. 15819726

Brand: Applied Biosystems™ 401956

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

These are septa covers for 0.5mL sample tubes, for use with the 310 Genetic Analyzer.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Each package contains 500 septa.
For Use With (Equipment) 310 Genetic Analyzer
For Use With (Application) Sequencing
Product Type Septa
Quantity 500 Each
Shipping Condition Room Temperature
Tube Type Disposable Tube, Conical Tube, Assay Tube

For research use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.