missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Septa Cathode Buffer Container, for 3500 and SeqStudio™ Flex
Shop All Applied Biosystems Products
Click to view available options
Quantity:
10 Containers
Unit Size:
Pack of 10
Description
These septa are for both compartments of the Cathode Buffer Container and are for use with 3500 and SeqStudio Flex series genetic analyzers.
Specifications
Specifications
| Content And Storage | This product contains 10 septa. |
| For Use With (Equipment) | 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers |
| Product Type | Septa Cathode Buffer Container |
| Quantity | 10 Containers |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction