missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Septa Cathode Buffer Container Dx

Product Code. 15978826 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
10 Ea.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15978826 10 Ea. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15978826 Supplier Applied Biosystems™ Supplier No. 4410716
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This is a septa cathode buffer container intended for use with Applied Biosystems Dx genetic analyzers.

This is a septa cathode buffer container intended for use with Applied Biosystems Dx genetic analyzers.

This is a septa cathode buffer container intended for use with the 3500 Dx/3500xL Dx Genetic Analyzer.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 Ea.
For Use With (Equipment) 3500 xL Dx Genetic Analyzer, 3500 Dx Genetic Analyzer
Product Type Septa Cathode Buffer Container
Shipping Condition Room Temperature
Content And Storage Each Pk. contains 10 containers.

For In Vitro Diagnostic Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.