missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Septa for 8-425 Screw Thread Caps

Product Code. 12470128
Click to view available options
Material:
PTFE, Silicone
Unit Size:
Pack of 100
Pack of 1000
This item is not returnable. View return policy

Product Code. 12470128

Brand: Thermo Scientific™ C401355

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Thermo Scientific™ National Septa for 8-425 Screw Thread Caps are constructed of PTFE, PTFE/rubber, or PTFE/silicone to suit a variety of applications.

Thermo Scientific™ National Septa for 8-425 Screw Thread Caps are constructed of PTFE, PTFE/rubber, or PTFE/silicone to suit a variety of applications.

  • PTFE Disk septa are chemically inert
  • PFTE red rubber septa are economical for many GC applications
  • PTFE/silicone septa are most popular for HPLC and IC applications
  • Pre-slit septa are easier to pierce with thin gauge needles
TRUSTED_SUSTAINABILITY

Specifications

Color Clear, Red
Diameter (Metric) Outer 8 mm
Material PTFE, Silicone
Septum PTFE/Silicone
Closure Size 8-425
For Use With (Equipment) Hitachi Autosamplers
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.