missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ 96-Well Septa Dx

Product Code. 16291041 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
20 Pk.
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16291041 20 Pk. Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16291041 Supplier Applied Biosystems™ Supplier No. 4410700
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This is a 96-well septa designed for use with Applied Biosystems Dx genetic analyzers.

This is a 96-well septa designed for use with Applied Biosystems Dx genetic analyzers.

Note: Applied Biosystems Dx genetic analyzers and consumables are for distribution and use in select countries only.

This is a 96-well septa designed for use with the 3500 Dx/3500xL Dx Genetic Analyzer.

Septa and plate base/retainer kits for 8 tube strip, 96 and 384-well plates used with the 3500 Dx Series Genetic Analyzers.

Note: The 3500 Dx Series Systems and consumables are for distribution and use in select countries only.

TRUSTED_SUSTAINABILITY

Specifications

Compatibility 96-Well Microtiter Plates, MicroAmp™ Optical 96-Well Reaction Plates
Quantity 20 Pk.
Format 96-well
For Use With (Equipment) 3500 xL Dx Genetic Analyzer, 3500 Dx Genetic Analyzer, 3500 xL Genetic Analyzer, 3500 Genetic Analyzer
Product Type Septa
Shipping Condition Room Temperature
Content And Storage Ambient storage.
Each Pk. contains 20 sets.

For In Vitro Diagnostic Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.