missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SEPT6 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA519024
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine and canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.
SEPT6 is a member of the septin family of GTPases. Members of this family are required for cytokinesis. This gene is a member of the septin family of GTPases. Members of this family are required for cytokinesis. One version of pediatric acute myeloid leukemia is the result of a reciprocal translocation between chromosomes 11 and X, with the breakpoint associated with the genes encoding the mixed-lineage leukemia and septin 2 proteins. This gene encodes four transcript variants encoding three distinct isoforms. An additional transcript variant has been identified, but its biological validity has not been determined.
Specifications
| SEPT6 | |
| Polyclonal | |
| Unconjugated | |
| SEPTIN6 | |
| 2810035H17Rik; 2-Sep; 6-Sep; C920001C06Rik; hypothetical protein LOC540783; Kiaa0128; MGC16619; MGC20339; mKIAA0128; RP5-876A24.2; SEP2; Sep6; SEPT11; SEPT2; SEPT6; septin 11; septin 2; septin 6; SEPTIN6; septin-6 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 23157 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q14141 | |
| SEPTIN6 | |
| Synthetic peptide sequence (DEVNAFKQRKTA) corresponding to the internal amino acids of 38961 (aa 391-403). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction