Learn More
SEPT6 Mouse, anti-Human, CoraLite Plus 488, Clone: 1B1G1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-68173-20UL
Description
The SEPT6 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SEPT6. This antibody recognizes human, mouse, pig, rabbit, rat antigen. The SEPT6 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| SEPT6 | |
| 1B1G1 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human, Mouse, Pig, Rabbit, Rat | |
| 155 kDa | |
| IgG1 |
| Immunofluorescence | |
| 1000 μg/mL | |
| KIAA0128, SEP2, SEPT2, SEPT6, septin 6 | |
| 20 μL | |
| 23157 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |