missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SEPT12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569861
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Yeast: 92%; Zebrafish: 86%.
Septins, such as SEPT12, are conserved GTP-binding proteins that function as dynamic, regulatable scaffolds for the recruitment of other proteins. They are involved in membrane dynamics, vesicle trafficking, apoptosis, and cytoskeleton remodeling, as well as infection, neurodegeneration, and neoplasia (Hall et al., 2005 [PubMed 15915442]).
Specifications
| SEPT12 | |
| Polyclonal | |
| Unconjugated | |
| SEPT12 | |
| 1700028G04Rik; 4933413B09Rik; SEPT12; septin 12; SEPTIN12; septin-12; SPGF10; testicular tissue protein Li 168 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 71089 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| 0 | |
| SEPT12 | |
| synthetic peptide directed towards the middle region of human SEPT12. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction