Learn More
SEPT11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14672-1-AP-20UL
 View more versions of this product
Product Code. 16883424
  • £135.00 / 20µL
Description
SEPT11 belongs to the conserved septin family of filament-forming cytoskeletal GTPases that are involved in a variety of cellular functions including cytokinesis and vesicle trafficking.
Specifications
Specifications
SEPT11
Polyclonal
Unconjugated
SEPT11
SEPT11, septin 11
Rabbit
Antigen Affinity Chromatography
RUO
305227, 52398, 55752
-20°C
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, Western Blot
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
B3GNI6, Q8C1B7, Q9NVA2
SEPT11
SEPT11 Fusion Protein Ag6342
20 μL
Primary
Human, Rat, Mouse