Learn More
STEMCELL Technologies SepMate™-50 (IVD)
Product Code. 15654468
Encompass
Click to view available options
Quantity:
100 Tubes
Unit Size:
Pack of 100
Product Code. 15654468
Brand: STEMCELL Technologies undefined
Only null left
Add to basket
Add to basket
50 mL centrifuge tube with an insert for density gradient centrifugation
  • Simplify peripheral blood mononuclear cell (PBMC) isolation by incorporating SepMate into your density gradient centrifugation step
  • SepMate tubes contain an insert that creates a barrier between the density gradient medium and blood, thus eliminating the need for careful blood layering and allowing mononuclear cells to be easily harvested with a simple pour
  • This product can be used with RosetteSep to isolate specific immune cell subsets
  • SepMate is manufactured under cGMP and registered as an In Vitro Diagnostic (IVD) device in Australia, Canada, Europe, and the USA
  • In China, SepMate is considered a nonmedical device by the China Food and Drug Administration (CFDA) and should be used as general lab equipment
  • The end user is responsible for determining whether the product is suitable for their specific application
  • Users in all other countries, please refer to SepMate-15 (RUO) (Catalog #86415) and SepMate-50 (RUO) (Catalog #86450)
Specifications
For Use With Cell Isolation; In Vitro Diagnostic; B Cells; Dendritic Cells; Monocytes; Mononuclear Cells; NK Cells; T Cells; T Cells, CD4+; T Cells, CD8+; T Cells, Other Subsets; T Cells, Regulatory
Quantity 100 Tubes