missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEPHS2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77776
This item is not returnable.
View return policy
Description
SEPHS2 Polyclonal specifically detects SEPHS2 in Mouse samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Knockdown Validated.
Specifications
| SEPHS2 | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| EC 2.7.9.3, Selenophosphate synthase 2, selenophosphate synthetase 2, SPS2water dikinase 2 | |
| Rabbit | |
| 47.68 kDa | |
| 0.1 mg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 22928 | |
| Mouse | |
| Purified |
| Western Blot, ELISA, Immunoprecipitation | |
| 2.3 mg/ml | |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:20000, Immunoprecipitation 1:100 | |
| AAC53024, P97364 | |
| SEPHS2 | |
| This Protein A purified SEPHS2 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a full-length recombinant protein corresponding to mouse SEPHS2. (Uniprot: P97364) | |
| Protein A purified | |
| RUO | |
| Primary | |
| This antibody reacts with mouse SEPHS2 and shows partial cross-reactivity with SPS1. | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction