missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEPHS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16635-1-AP-150UL
This item is not returnable.
View return policy
Description
This protein encodes an enzyme that synthesizes selenophosphate from selenide and ATP. Selenophosphate is the selenium donor used to synthesize selenocysteine, which is co-translationally incorporated into selenoproteins at in-frame UGA codons.Specifications
| SEPHS1 | |
| Polyclonal | |
| Unconjugated | |
| SEPHS1 | |
| SELD, Selenide, water dikinase 1, Selenium donor protein 1, Selenophosphate synthase 1, selenophosphate synthetase 1, SEPHS1, SPS, SPS1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 109079, 22929, 291314 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P49903, Q8BH69 | |
| Sephs1 | |
| SEPHS1 Fusion Protein Ag9978 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction