missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEPHS1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-32312
This item is not returnable.
View return policy
Description
SEPHS1 Polyclonal specifically detects SEPHS1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SEPHS1 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| SEPHS1 | |
| Recombinant protein encompassing a sequence within the center region of human Selenophosphate synthetase 1. The exact sequence is proprietary. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 22929 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.92mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| EC 2.7.9.3, MGC4980, SELD, Selenium donor protein 1, Selenophosphate synthase 1, selenophosphate synthetase 1, selenophosphate synthetase 1 +E9a, selenophosphate synthetase 1 delta E2, selenophosphate synthetase 1 delta E8, selenophosphate synthetase 1 major, SPS1selenophosphate synthetase 1 +E9, water dikinase 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Tested in human. Predicted cross-reactivity based on sequence identity: Rhesus Monkey (100%) and Chimpanzee (100%). Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction