missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEPHS1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38270-100ul
This item is not returnable.
View return policy
Description
SEPHS1 Polyclonal antibody specifically detects SEPHS1 in Human,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| SEPHS1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10 - 1:100, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| EC 2.7.9.3, MGC4980, SELD, Selenium donor protein 1, Selenophosphate synthase 1, selenophosphate synthetase 1, selenophosphate synthetase 1 +E9a, selenophosphate synthetase 1 delta E2, selenophosphate synthetase 1 delta E8, selenophosphate synthetase 1 major, SPS1selenophosphate synthetase 1 +E9, water dikinase 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-392 of human SEPHS1 (NP_036379.2).,, Sequence:, MSTRESFNPESYELDKSFRLTRFTELKGTGCKVPQDVLQKLLESLQENHFQEDEQFLGAVMPRLGIGMDTCVIPLRHGGLSLVQTTDYIYPIVDDPYMMGRIACANVLSDLYAMGVTECDNMLMLLGVSNKMTDRERDKVMPLIIQGFKDAAEEAGTSVTGGQTVLNPWIVLGGVATTVCQPNEFIMPDNAVPGDVLVLTKPLGTQVAVAVHQWLDIPEKWNKIKLVVTQEDVELAYQEAMMNMARLNRTAAGLMHTFNAHAATDITGFGILGHAQNLAKQQRNEVSFVIHNLPVLAKMAAVSKACGNMFGLMHGTCPETSGGLLICLPREQAARFCAEIKSPKYGEGHQAWIIGIVEKGNRTARIIDKPRIIEVAPQVATQNVNPTPGATS | |
| 100 μL | |
| Stem Cell Markers | |
| 22929 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction