missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sepharose Agarose Gel Filtration Media
Shop All Cytiva Products
Click to view available options
Flow Rate:
<10 cm/hr.
<11.5 cm/hr.
<14 cm/hr.
<15 cm/hr.
<26 cm/hr.
<30 cm/hr.
Particle Size:
45 to 165 μm
60 to 200 μm
pH Range:
3 to 12
3 to 13
4 to 9
Support Type:
2% Agarose
4% Agarose
6% Agarose
Unit Size:
1 L
Description
European Pharmacopoeia Solutions
- Reagents and Standard solutions in Chapters 2 and 4 of the current Ph. Eur.
- All are manufactured and tested in compliance with the Ph. Eur.
- Supplied with a Certificates of Analysis
- Lot No. and expiry date are stated on all product labels
Order Info
Supplied in 1L plastic containers
Specifications
Specifications
| Quantity | 1 L |
| Flow Rate | <15 cm/hr. |
| Particle Size | 60 to 200 μm |
| Product Type | Sepharose Agarose Gel Filtration Media |
| Format | Solid Bead |
| Diameter (Metric) Inner | 5cm |
| For Use With (Application) | Allow characterization or cleaning up of samples containing components of diverse molecular weight |
| Height (English) | 3.94 in. |
| Height (Metric) | 10cm |
| Max. Pressure | 60-120 cm/h, pressure drop cm H2O/bed height=15, bed height 10 cm, 5 cm i.d. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction