missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sephadex™ LH-20

Product Code. 11530784 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11530784 500 g 500g
10309794 25 g 25g
11520784 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11530784 Supplier Cytiva Supplier No. 17009002
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use for molecular sizing of natural product such as steroid, terpenoid, lipid, low molecular weight peptide. Cytiva Life Sciences™Sephadex™ LH-20 is a liquid chromatography media useful to analytical and industrial scale for preparation of closely related molecular species.

  • Prepared for use in polar organic solvents and aqueous solvent mixtures
  • Made from hydroxypropylated dextran beads that have been crosslinked to yield a polysaccharide network
  • Hydrophilic and lipophilic dual nature of the matrix enables powerful chromatographic selectivity
  • Batch-to-batch reproducibility
  • Partical size dry: 18μ-111μm
  • Fractionation range globular proteins: 4x103 to 5 x 103

  • Stable in most aqueous and organic eluent systems
  • Shelf life: 5years
  • Autoclavable: 20 min at 121°C
  • Flow Velocity <∽12cm/min
  • pH stability working range: 2-13

Analytical and industrial scales for the preparation of closely related molecular species

TRUSTED_SUSTAINABILITY

Specifications

Quantity 500 g
Flow Rate 720 cm/hr.
Particle Size 18 to 111 μm in dry form
Product Type Sephadex LH-20 Media
Format Solid Spherical Bead
Diameter (Metric) Inner 2.6cm
For Use With (Application) For fractionation of lipids, hormones, vitamins, and other small biomolecules in polar organic solvents and aqueous solvent mixtures
Height (English) 11.81 in.
Height (Metric) 30cm
Max. Pressure 25-45 cm/h, pressure drop cm H2O/bed height=2, beh height 30 cm, 2.6 cm i.d.
pH Range 2 to 13
Support Type Hydroxypropylated, cross-linked Dextran
Sample Type Steroid, Terpenoid, Lipid, low molecular weight Peptide
Content And Storage 4 to 30°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.