missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sephadex™ G50 Fine Media

Suitable for gel filtration

Brand:  Cytiva 17-0042-02

Code : GE

Additional Details : Weight : 0.57900kg

 View more versions of this product

Product Code. 11544875

  • £1499.00 / 500g
Estimated Shipment: 22-05-2024
to see stock.
Add to basket

Description

Description

  • Particle size range 40 to 160μm

Specifications

Specifications

500g
34 to 208 μm in wet form, 20 to 80 μm in dry form
Sephadex G50 Media
5cm
3.93 in.
min 150 cm/h, bed height 10 cm, column 5 cm i.d.
Cross-linked Dextran
4 to 30°C
< Darcy´s Law
9 to 11mg/g
Solid Bead
For the separation of molecules Mr >30 000 from molecules Mr <1500 such as labeled protein or DNA from unconjugated dyesFor removeing small nucleotides from longer chain nucleic acids
10cm
2 to 10
DNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers