missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sephadex™ G15 Media
Shop All Cytiva Products
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
- For purposes of determining maximum flowrate and maximum pressure, beads behave as rigid spheres according to Darcy's Law
- Working pH range 2 to 13
- Particle size range for wet beads 60 to 180μm
- Bed volume in distilled water 2.5 to 3.5mL/g
- Exclusion limit: <1.5 x 103 for dextrans, <1.5 x 103 for globular proteins
Specifications
Specifications
| Quantity | 100 g |
| Flow Rate | < Darcy´s Law |
| Particle Size | 60 to 180 μm in wet form, 40 to 120 μm in dry form |
| Bed Volume | 2.5 to 3.5mL/g in Distilled Water |
| Product Type | Sephadex G15 Media |
| Format | Solid Bead |
| Diameter (Metric) Inner | 2.6cm |
| For Use With (Application) | Useful for desalting and Buffer exchange of Peptides and small biomolecules >1500 M.W. |
| Height (English) | 11.81 in. |
| Height (Metric) | 30cm |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction