missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sephadex™ G10 Media

Product Code. 11594865 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11594865 500 g 500g
11584865 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11594865 Supplier Cytiva Supplier No. 17001002
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gel filtration medium for desalting and buffer exchange of peptides and small biomolecules

  • Beads behave as rigid spheres obeying Darcy's Law for purposes of determining maximum flowrate and maximum operating pressure
  • Working pH range 2 to 13
  • Particle size range 55 to 165μm
  • Bed volume in distilled water 2-3mL/g
  • Exclusionation limit: 7 x 102 dextrans, 7 x 102 globular proteins

TRUSTED_SUSTAINABILITY

Specifications

Quantity 500 g
Flow Rate < Darcy´s Law
Particle Size 55 to 165 μm in wet form, 40 to 120 μm in dry form
Bed Volume 2 to 3mL/g in Distilled Water
Product Type Sephadex G10 Media
Format Solid Bead
Diameter (Metric) Inner 2.6cm
For Use With (Application) Desalting and Buffer exchange of Peptides and small biomolecules >700 M.W.
Height (English) 11.81 in.
Height (Metric) 30cm
Max. Pressure 24-49 cm/h, pressure drop cm H2O/bed height = 2, bed height 30 cm, 2.6 cm i.d
pH Range 2 to 13
Support Type Cross-linked Dextran
Sample Type Peptides, Small Biomolecules
Content And Storage 4 to 30°C
Show More Show Less
Product Title
Select an issue

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.