missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sephadex™ G-25 Media

Product Code. 11550774 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
100 g
500 g
Flow Rate:
>100 cm/hr.
>11 cm/hr.
>47 cm/hr.
>480 cm/hr.
Particle Size:
15 to 88 μm (Wet Form), >20 μm (Dry Form)
17 to 132 μm (Wet Form), >20 μm (Dry Form)
38 to 235 μm (Wet Form), >50 μm (Dry Form)
87 to 510 μm (Wet Form), >100 μm (Dry Form)
Unit Size:
100g
500g
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Flow Rate Particle Size unitSize
11550774 100 g >11 cm/hr. 15 to 88 μm (Wet Form), >20 μm (Dry Form) 100g
11570774 500 g >47 cm/hr. 17 to 132 μm (Wet Form), >20 μm (Dry Form) 500g
11510784 500 g >480 cm/hr. 87 to 510 μm (Wet Form), >100 μm (Dry Form) 500g
11560774 100 g >47 cm/hr. 17 to 132 μm (Wet Form), >20 μm (Dry Form) 100g
11500784 100 g >480 cm/hr. 87 to 510 μm (Wet Form), >100 μm (Dry Form) 100g
10669455 500 g >100 cm/hr. 38 to 235 μm (Wet Form), >50 μm (Dry Form) 500g
11580774 100 g >100 cm/hr. 38 to 235 μm (Wet Form), >50 μm (Dry Form) 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 11550774 Supplier Cytiva Supplier No. 17003101
Powering Proteomics
Use promo code 28003
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Use of Sephadex™ is well established as a gel filtration medium for desalting and buffer exchange in industrial applications. Cytiva Life Sciences™ Sephadex™ G-25 Media quickly desalts, removes contaminants and transfers to a new buffer in a single step.

Established medium for desalting and buffer exchange

  • Gel filtration medium
  • Available in coarse, medium, fine and superfine grades
  • All grades satisfy process-scale chromatography requirements
  • Available in lab packs or prepacked columns

Specifications:

  • Working pH range: 2 to 13
  • Bed volume (in distilled water): 4 to 6mL/g
  • Fractionation range dextrans/globular proteins: 1× 102 to 5 × 103 / 1 × 103 to 5 × 103 Mr
  • Max. flowrate/operating pressure: the beads behave as rigid spheres obeying Darcy's Law

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 g
Flow Rate >11 cm/hr.
Particle Size 15 to 88 μm (Wet Form), >20 μm (Dry Form)
Bed Volume 4 to 6mL/g in Distilled Water
Product Type Sephadex G-25 Media
Format Solid Spherical Bead
Diameter (Metric) Inner 2.6cm
For Use With (Application) Desalting and Buffer exchange and removal free label in industrial applications
Height (English) 11.81 in.
Height (Metric) 30cm
Max. Pressure >11 cm/h, pressure drop cm H2O/bed height=2, bed height 30 cm, 2.6 cm i.d.
pH Range 2 to 13
Support Type Cross-linked Dextran
Content And Storage 4 to 30°C, dry resins. Used resins 4 to 8 °C in 20% Ethanol or 0.01 M NaOH
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.