Learn More
Cytiva Sephadex™ G-25 Media
Use of Sephadex™ is well established as a gel filtration medium for desalting and buffer exchange in industrial applications. Cytiva Life Sciences™ Sephadex™ G-25 Media quickly desalts, removes contaminants and transfers to a new buffer in a single step.
Brand: Cytiva 17-0034-01
Code : GE
Additional Details : Weight : 0.12300kg
Description
Established medium for desalting and buffer exchange
- Gel filtration medium
- Available in coarse, medium, fine and superfine grades
- All grades satisfy process-scale chromatography requirements
- Available in lab packs or prepacked columns
Specifications:
- Working pH range: 2 to 13
- Bed volume (in distilled water): 4 to 6mL/g
- Fractionation range dextrans/globular proteins: 1× 102 to 5 × 103 / 1 × 103 to 5 × 103 Mr
- Max. flowrate/operating pressure: the beads behave as rigid spheres obeying Darcy's Law
Specifications
100g | |
87 to 510 μm in wet form, >100 μm in dry form | |
Sephadex G-25 Media | |
2.6cm | |
11.81 in. | |
>480 cm/h, pressure drop cm H2O/bed height=2, bed height 30 cm, 2.6 cm i.d. | |
Cross-linked Dextran |
>480 cm/hr. | |
4 to 6mL/g in Distilled Water | |
Solid Spherical Bead | |
Desalting and Buffer exchange and removal free label in industrial applications | |
30cm | |
2 to 13 | |
4 to 30°C, dry resins. Used resins 4 to 8 °C in 20% Ethanol or 0.01 M NaOH |