missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Sephadex™ G-100, MP Biomedicals™

Product Code. 11465222 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11465222 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11465222 Supplier MP Biomedicals™ Supplier No. 0219525550
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used in protein, affinity and gel filtration chromatography. Sephadex™ G-100, MP Biomedicals is a beaded gel filtration medium prepared by cross-linking dextran with epichlorohydrin under alkaline conditions. Its main application is group separation

Desalting with Sephadex is superior to dialysis because of the considerable time savings, the low dilution factor (which can be as low as 1.4 : 1), and the high activity recoveries, even with very small amounts of sample. Related applications include buffer exchange and the removal of small molecules during the preparation of large biomolecules, such as ampholytes, detergents, radioactive or fluorescent labels, and phenol (during DNA purification).

Specifications

Quantity 50 g
Flow Rate 40 cm/hr.
Particle Size 40 to 120 μm
Product Type Sephadex G-100 Media
Content And Storage Store at Room Temperature (15–30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.