missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sephadex™ G-100 Media

Product Code. 11544885 Shop All Cytiva Products
Click to view available options
Flow Rate:
1 mL/min.
4.2 mL/min.
Particle Size:
100 to 310 μm (Wet Form), 100 to 310 μm (Dry Form)
25 to 100 μm (Wet Form), 10 to 40 μm (Dry Form)
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 11544885

Brand: Cytiva 17006101

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Commonly used for desalting and buffer exchange of very large biomolecules

  • Available in two particle size ranges

Fine particle size

  • Particle size, dry: 40 to 120μm
  • Fractionation range, globular proteins: 4000 to 150,000
Superfine particle size
  • Particle size, dry: 10 to 40μm
  • Fractionation range, globular proteins: 4000 to 100,000

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 g
Flow Rate 1 mL/min.
Particle Size 25 to 100 μm (Wet Form), 10 to 40 μm (Dry Form)
Bed Volume 15 to 20mL/g in Distilled Water
Product Type Sephadex G-100 Media
Format Solid Bead
For Use With (Application) Useful for desalting and Buffer exchange of very large biomolecules
Max. Pressure 0.096 bar
pH Range 2 to 10
Support Type Cross-linked Dextran
Sample Type Large biomolecules
Content And Storage 4° to 30°C
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.