missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sephacryl™ S-200 High Resolution Media

Product Code. 11570784 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
750mL
Unit Size:
750mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11570784 750mL 750mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11570784 Supplier Cytiva Supplier No. 17058401
Powering Proteomics
Use promo code 28003
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Hydrophilic, rigid allyl dextran/bisacrylamide matrix for fast and reproducible purification of proteins, polysaccharides and other macromolecules

Chromatography media offer good recovery and low nonspecific adsorption. Enable gel filtration at laboratory and industrial scale, and may be purchased in prepacked columns.

  • For purifying antibodies, and serum and mid-size proteins
  • Excellent purification over a wide molecular weight range
  • High reproducibility due to long-term chemical and physical stability
  • High flow rates and recoveries
  • Well-suited to industrial-scale use
  • Storage conditions: 20% ethanol; 4° to 30°C
  • Available in a variety of selectivities and volumes

Chemical Stability (stable in all common buffers):

  • 1M acetic acid, 8M urea
  • 6M guanidine-HCl, 1% SDS
  • 2M NaCl, 24% ethanol
  • 30% propanol, 30% acetonitrile
  • 0.5M NaOH (for cleaning)
pH Stability
  • 3 to 11 (long-term and working)
  • 2 to 13 (short-term)
Physical Stability:
  • Negligible volume variation due to changes in pH or ionic strength
  • Autoclavability: At 121°C, pH 7 for 30 minutes (gel only)
  • Antimicrobial agent: 20% ethanol

TRUSTED_SUSTAINABILITY

Specifications

Quantity 750mL
Flow Rate >150 cm/hr.
Particle Size 25 to 75 μm
Bed Volume 19 to 230L
Product Type Sephacryl HR Media
Format Solid Spherical Bead
Diameter (Metric) Inner 40 to 140cm
For Use With (Application) Allow fast and reproducible purification of antibodies, serum proteins and mid-size proteins by gel filtration at laboratory and industrial scale
Height (English) 5.9 in.
Height (Metric) 15cm
Max. Pressure 100kPa
pH Range 3 to 11
Support Type Allyl Dextran and N, N'-methylenebisacrylamide
Sample Type Antibodies, serum and mid-size proteins
Content And Storage 4° to 30°C
Show More Show Less
compliance-icons
Product Identifier
  • Sephacryl S-200 High Resolution, 750 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.