missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEP 15. Rabbit anti-Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | SEP 15. |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
44454 Polyclonal specifically detects 44454 in Rat samples. It is validated for Western Blot.Specifications
| SEP 15. | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Rat | |
| 15 kDa selenoprotein | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of Rat sep15 (NP_579831). Peptide sequence LFRGLQIKYVRGSDPVLKLLDDNGNIAEELSILKWNTDSVEEFLSEKLER | |
| Affinity purified |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS buffer, 2% sucrose | |
| 9403 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title