missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SenTraGor™

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
SenTraGor™ is a reagent for detecting lipofuscin in senescent cells. A highly lipophilic and biotinylated analog of Sudan Black B; can be detected using an anti-biotin antibody. Can be used in vivo and in vitro in any biological material including cell cultures, fresh and frozen tissues, and fixed embedded samples. Compatible with immunohistochemistry, flow cytometry and immunofluorescence analysis.
Specifications
Specifications
| Chemical Name or Material | (2-Methyl-6-((E)-(4-((E)-phenyldiazenyl)naphthalen-1-yl)diazenyl)-2,3-dihydro-1H-perimidin-2-yl)methyl 5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoate |
| CAS | 2171036-89-6 |
| Quantity | 10 mg |
| Target | Buffers, Chelators and Reagents |
| Molecular Formula | C39H38N8O3S |
| Purity | 0.95 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction