missing translation for 'onlineSavingsMsg'
Learn More

WTW™ SenTix™ 42 pH Electrode
GREENER_CHOICE

Product Code. 10101441
Change view
Click to view available options
Connector Types:
BNC, Banana Plug
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types unitSize
10101441 BNC, Banana Plug Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10101441 Supplier WTW™ Supplier No. 103637
Add to basket
Add to basket
This item is not returnable. View return policy

A standard pH gel electrode, ideal for portable measurements and in-lab work across a range of applications.

  • Maintenance-free gel reference system across the entire product line
  • Quick response time by good connection to the medium
  • Best results thanks to high-quality membrane glasses
  • Excellent where reliable measured values are needed even under the most adverse conditions
  • Especially economical, only need minimal cleaning
  • With temperature sensor and BNC cable
TRUSTED_SUSTAINABILITY

Specifications

Connector Types BNC, Banana Plug
Junction Fiber
Material Plastic Shaft, Fiber Diaphragm
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 80°C
Type SenTix™ pH Electrode
For Use With (Equipment) Vario pH meter
Includes BNC, banana electrode plug, 1m cable
Model SenTix™ 42
Probe 30kilohms Built-in NTC
Storage Requirements Immerse the combination electrode in reference electrolyte (KCl 3mol/L, Ag+ free), pH combination electrodes must not be stored dry or in deionized water
Temperature (Metric) Range 0°C to 80°C
Length (Metric) Cable 1 m
Length (Metric) Stem 120 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.