Learn More
Mettler Toledo™ Sensor Holder, EasyPlace, Triple
Product Code. 17591016 Shop All Mettler Toledo Products
Change view
Click to view available options
For Use With (Equipment):
EasyPlace
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
17591016 EasyPlace Each
1 options
Product Code. 17591016
Brand: Mettler Toledo™ 30671575
Only null left
Add to basket
Add to basket
Sensor holder for the EasyPlace electrode arm. Fits up to three sensors.
  • Three-in-One
  • For multi-channel measurements, external references or temperature sensors - position multiple sensor with one EasyPlace
TRUSTED_SUSTAINABILITY
Specifications
Type Sensor Holder, EasyPlace Triple
For Use With (Equipment) EasyPlace