Learn More
WTW™ SensoLyt™ ORP 900-P
GREENER_CHOICE
Product Code. 15364912
Change view
Click to view available options
For Use With (Equipment):
MPP IDS or AS/IDS-x
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
15364912 MPP IDS or AS/IDS-x Each
1 options
Product Code. 15364912 Supplier WTW™ Supplier No. 103749
Only null left
Add to basket
Add to basket
ORP electrodes deliver a sensitive voltage signal from reactions in waterous solutions, where the reaction partners exchange loads on the Pt metal surface of the electrode
ORP electrodes deliver a sensitive voltage signal from reactions in waterous solutions, where the reaction partners exchange loads on the Pt metal surface of the electrode. The scale is comparable with that of pH measurement. Typical areas of use are the monitoring of the disinfection effect, the determination of ORP potentials in biochemical reactions, measuring in waters of different quality and more.This is exactly where the concept of IDS sensors comes in. Digital data transfer is protected from interference, allows the use of long cables and permits important additional information, such as calibration data and sensor characteristics.WTW's proven ORP electrodes are now available as sustainable plug head versions. The universal plug head fits the sensor with wireless functionality– disturbing cables are no longer required.Furthermore it can be connected to AS/IDS-x cables with lengths of up to 100 m. With this new technology WTW significantly expands the range of applications and the measuring comfort of its ORP electrodes.
TRUSTED_SUSTAINABILITY
Specifications
Type Combined IDS pH Electrode
Includes Polymer electrolyte and glass shaft, integrated temperature sensor, Electrode with waterproof plug head for connection to cable AS / IDS-x or MPP IDS.
For Use With (Equipment) MPP IDS or AS/IDS-x