missing translation for 'onlineSavingsMsg'
Learn More

Sensititre™ YO10

Product Code. 15397137
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15397137 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15397137 Supplier Thermo Scientific Oxoid Supplier No. YYO10
  • ONLINE EXCLUSIVE PRICE
    £455.00
    £446.10 / Pack of 10
    Save £8.90
    1% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Simplify yeast testing for Candida sp. and reduce cost per test using this plate with nine antifungals, including Anidulafungin and Micafungin

Simplify yeast testing with the Thermo Scientific™ Sensititre™ YeastOne™ YO10 AST Plate with nine antifungals, including Anidulafungin and Micafungin. Colorimetric Thermo Scientific™ alamarBlue™ agent provides reliable, easy and consistent endpoint determination, with visual read option, for efficient workflow and accurate results.

Simplify yeast testing with the Sensititre YeastOne YO10 AST Plate with nine antifungals, including Anidulafungin and Micafungin. 

  • Room-temperature storage eliminates inventory control concerns
  • Individual packaging allows laboratory to test one plate at a time with no waste
  • Inclusive on-scale QC ranges provide immediate quality assurance of testing methodology
  • 24-hour incubation time enables timely results for clinicians

Not all products are available for sale in all territories. Please inquire.

VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Antimicrobial Agents Various
Broth Type YeastOne Broth (Part No. Y3462)
Description Sensititre YeastOne Plate w/ Anidulafungin and Micafungin
Antibiotic Code Various
Product Type AST Plate
Target Organism Class Yeast
Test Type Minimum Inhibitory Concentration (MIC)
For Use With (Application) Candida sp.
Incubation Conditions 35°C O2
Incubation Time 24-25 hr
Isolates Per Plate 1
Packaging Type Box
Quantity 10/Pk.
Reading Options Manual
Specimen Type Candida Species
Transfer Volume 20μL
Volume Per Well 100 μL
Species Candida species
Show More Show Less

CE/IVD-marked.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.