missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sensititre™ STP6F

Product Code. 17738574
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17738574 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17738574 Supplier Thermo Scientific™ Supplier No. YSTP6F
  • ONLINE EXCLUSIVE PRICE
    £302.00
    £128.95 / Pack of 10
    Save £173.05
    57% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Perform susceptibility testing of Streptococcus species with this FDA cleared, convenient plate including Tigecycline.

Perform susceptibility testing of Streptococcus species with Thermo Scientific™ Sensititre™ Streptococcus STP6F AST Plate. This plate is FDA-cleared for automated or manual reading of Streptococcus species using the Thermo Scientific™ Sensititre™ ARIS™ 2X ID/AST System, Thermo Scientific™ Sensititre™ OptiRead System, or Sensititre™ manual platforms, including the Thermo Scientific™ Sensititre™ Vizion™ System for maximum versatility.

Eliminate offline tests with a full range of antimicrobials, including Tigecycline, with the Sensititre Streptococcus STP6F AST Plate. 

  • Individual packaging allows laboratories to test one isolate at a time
  • Test 20 antimicrobials on a single AST plate for cost-effective results
  • True MIC results provide better resistance tracking
  • Extended shelf life and room-temperature storage eliminate inventory control concerns

Not all products are available for sale in all territories. Please inquire.

VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

0.5 McFarland Standard MHB (Part No. T346205)
Antimicrobial Agents Various
Broth Type MHB w/ LHB (Part No. CP114)
Description Sensititre Streptococcus species MIC Plate
Antibiotic Code Various
Product Type AST Plate
Target Organism Class Fastidious Organisms
Test Type Minimum Inhibitory Concentration (MIC)
For Use With (Application) All Streptococcus sp.
Incubation Conditions 34-36°C O2
Incubation Time 20-24 hr
Isolates Per Plate 1
No. per Pack 10
Packaging Type Box
Quantity 10/Pk.
Reading Options Autoread or manual
Transfer Volume 100µL
Volume Per Well 100 μL
Species Streptococcus spp., Haemophilus influenzae
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.