Learn More
Thermo Scientific™ Sensititre™ Manual Viewbox
Product Code. 16209121
Change view
Click to view available options
Product Type:
Manual Viewbox
Unit Size:
Each
Product Code. Product Type Unit Size
16209121 Manual Viewbox Each
1 options
Product Code. 16209121 Supplier Thermo Scientific™ Supplier No. YV4007
  • ONLINE EXCLUSIVE PRICE
    £637.00
    £519.53 / Pack of 1
    Save £117.47
    18% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Perform simple visual reads of your 96-well microtitre plates with the mirrored Thermo Scientific™ Sensititre™ Manual Viewbox.
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Description Sensititre Manual Viewbox
Product Type Manual Viewbox
Test Type Minimum Inhibitory Concentration (MIC)