missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sensititre™ GPALL1F

Product Code. 17842876 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17842876 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17842876 Supplier Thermo Scientific™ Supplier No. GPALL1F
  • ONLINE EXCLUSIVE PRICE
    £267.31
    £111.69 / Pack of 10
    Save £155.62
    58% Off
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Perform accurate AST of non-fastidious Gram-positive isolates on a single platform with this convenient plate, featuring D-Test and Cefoxitin Screen.

Confidently perform susceptibility testing of non-fastidious Gram-positive isolates with Thermo Scientific™ Sensititre™ Gram Positive GPALL1F AST Plate, featuring D-Test and Cefoxitin Screen.

Test for non-fastidious Gram-positive isolates with true MIC results using Sensititre Gram Positive GPALL1F AST Plate, featuring D-Test and Cefoxitin Screen. 

  • The plate has fewer limitations for Enterococcus spp. and Staphylococcus spp. versus other automated systems
  • Full-range, true MIC readings provide results for better detection of resistance
  • Automated or manual read options are available for ultimate flexibility
  • 18 - 24-month shelf life and room-temperature storage simplify inventory control

Not all products are available for sale in all territories. Please inquire.

VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

0.5 McFarland Standard H2O (Part No. T3339)
Antimicrobial Agents Various
Broth Type MHB (Part No. T3462)
Description Sensititre Gram Positive All-in-One Plate
Antibiotic Code Various
Product Type AST Plate
Target Organism Class Non-fastidious Gram-positive Organisms
Test Type Minimum Inhibitory Concentration (MIC)
Incubation Conditions 35°C O2
Incubation Time 18-24 hr
Isolates Per Plate 1
No. per Pack 10
Quantity 10/Pk.
Reading Options Autoread or manual
Transfer Volume 10µL or 30µL* (*Aid in detection of resistance mechanisms for gram positive organisms; see pack insert)
Volume Per Well 50 μL
Species Non-fastidious Gram-positive Isolates
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.