missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sensititre™ GN6F

Product Code. 17869050
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17869050 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17869050 Supplier Thermo Scientific™ Supplier No. YGN6F
  • ONLINE EXCLUSIVE PRICE
    £264.00
    £186.07 / Pack of 10
    Save £77.93
    29% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Accurately perform ceftazidime/avibactam and ceftolozane/tazobactam susceptibility testing of Gram negative organisms by micro-broth dilution.

Confidently perform susceptibility testing with Gram negative antimicrobials, including ceftazidime/avibactam and ceftolozane/tazobactam using Thermo Scientific™ Sensititre™ Gram Negative GN6F AST Plates. Address your formulary needs with this FDA-cleared, micro-broth dilution plate for antimicrobial susceptibility testing which meets CLSI standards. Obtain true minimum inhibitory concentration (MIC) results to both inform targeted therapy decisions for resistant infections with otherwise limited or no treatment options and to support antimicrobial stewardship to preserve effectiveness.

Product Features

Expand treatment options by performing ceftazidime/avibactam and ceftolozane/tazobactam susceptibility testing of Gram-negative organisms with Sensititre Gram Negative GN6F AST Plates. Generate clinically relevant data, detect emerging resistance, and help guide timely and accurate therapy intervention by utilizing micro-broth dilution.

  • Accurate: meets CLSI and FDA breakpoints
  • Quantitative: precise MIC results aid informed patient therapy decisions
  • Efficient: accurate results the first time, thereby reducing unnecessary testing and protocols
  • Flexible: automated, semi-automated or manual read options are available for ultimate flexibility
  • Easy-to-store: extended shelf life and room-temperature storage simplify inventory control


The Sensititre System is a leader in AST solutions, offering flexible, customizable testing options for laboratories of all sizes.
 

  • Scalable: flexible instrumentation options designed to be modular in order to meet the unique needs of individual lab space, workload, and budget requirements
  • Flexible: over 240 antimicrobials available in extended dilution ranges, access to the newest antimicrobials, and the ability to consolidate AST onto one platform
  • Tailor-made: design your own AST panel to meet specific formulary requirements and prescription protocols and to clinicians that are based on real-time trends and local epidemiology

Not all products are available for sale in all territories. Please inquire.

VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

0.5 McFarland Standard H2O (Part No. T3339)
Antimicrobial Agents Various
AST Type MIC
Broth Type MHB (Part No. T3462)
Concentration Multiple
Description Sensititre Gram Negative GN6F Plate
Antibiotic Code Various
Format Multi-antibiotic Microtitre Panels
Product Type AST Plate
Target Organism Class Gram-negative Microorganisms
Test Type Minimum Inhibitory Concentration (MIC)
Incubation Conditions 35°C O2
Incubation Time 18 hr
Isolates Per Plate 1
No. per Pack 10
Packaging Type 10 x Microtitre Panels
Quantity 10/Pk.
Reading Options Autoread or manual
Transfer Volume For Enterobacteriaceae and Non-Enterobacteriaceae: 10μL, 30μL* (*Aid in detection of resistance mechanisms for gram negative organisms; see pack insert). For Proteus sp.: 1μL
Volume Per Well 50 μL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.