Learn More
Sensititre™ Gram Negative EUMDROF AST Plate
Product Code. 16496069
Change view
Click to view available options
Broth Type:
MHB (part no. T3462)
Unit Size:
Pack of 10
Product Code. Broth Type Unit Size
16496069 MHB (part no. T3462) Pack of 10
1 options
Product Code. 16496069 Supplier Thermo Scientific Oxoid Supplier No. YEUMDROF
  • ONLINE EXCLUSIVE PRICE
    £350.00
    £318.16 / Pack of 10
    Save £31.84
    9% Off
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Consolidate MDRO confirmation susceptibility testing onto a single plate that includes colistin, Ceftazidime/Avibactam and Ceftolozane/Tazobactam.
Perform susceptibility testing of non-fastidious Gram-Negative species isolates on a single platform with Thermo Scientific™ Sensititre™ Gram-Negative EUMDROF AST Plate.
The convenient, standard AST plate meets EUCAST breakpoints for relevant antimicrobials on the plate.
Consolidate MDRO confirmation susceptibility testing onto a single plate format that includes colistin, Ceftazidime/Avibactam and Ceftolozane/Tazobactam.
Achieve reliable, consistent antimicrobial susceptibility results for non-fastidious Gram-Negative isolates with the Sensititre Gram-Negative EUMDROF AST Plate. Generate clinically relevant data, detect emerging resistance, and help guide timely and accurate therapy intervention by utilizing micro-broth dilution.
  • Monitor susceptibility among Gram-negative isolates with full-range MIC values.
  • For ultimate flexibility automated or manual read options are available
  • Eliminate inventory control concerns with an extended shelf life (18 to 24 months) and room temperature storage.
 The Sensititre System is a leader in AST solutions, offering flexible, customizable testing options for laboratories of all sizes.
  • Scalable: flexible instrumentation options designed to be modular in order to meet the unique needs of individual lab space, workload, and budget requirements
  • Flexible: over 240 antimicrobials available in extended dilution ranges, access to the newest antimicrobials, and the ability to consolidate AST onto one platform
  • Tailor-made: design your own AST panel to meet specific formulary requirements and prescription protocols and to clinicians that are based on real-time trends and local epidemiology
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
0.5 McFarland Standard Demineralized Water (part no. T3339)
Antimicrobial Agents Various
Broth Type MHB (part no. T3462)
Description Sensititre Gram Negative EUMDROF AST Plate
Antibiotic Code Various
Product Type AST Plate
Target Organism Class Non-fastidious, Gram-negative Organisms
Test Type Minimum Inhibitory Concentration (MIC)
Incubation Conditions 35°C; O2
Incubation Time 18-24 hrs
Isolates Per Plate 1
Packaging Type Box
Quantity 10/Box
Reading Options Autoread or manual
Transfer Volume 10μL
Volume Per Well 50 μL