missing translation for 'onlineSavingsMsg'
Learn More

Sensititre™ Bovine/Porcine BOPO6F Vet AST Plate

Product Code. 15824947 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15824947 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15824947 Supplier Thermo Scientific Supplier No. YBOPO6F
  • ONLINE EXCLUSIVE PRICE
    £256.57
    £229.65 / Pack of 10
    Save £26.92
    10% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Perform accurate AST with a bovine/porcine-specific formulary containing food animal antimicrobials using this convenient plate.

Perform accurate AST with a bovine/porcine-specific formulary, containing food animal antimicrobials using the Thermo Scientific™ Sensititre™ Bovine/Porcine BOPO6F AST Plate. Sensititre Bovine/Porcine BOPO6F AST Plate delivers accurate results on a single plate format, reducing the need for offline tests and retests.

Achieve reliable, consistent results for non-fastidious Gram-positive and Gram-negative isolates of veterinary origin with Sensititre Bovine/Porcine BOPO6F Plate.

  • Extended shelf life, room temperature storage eliminates inventory control concerns.
  • Individual packaging allows the laboratory to test one plate at a time with no waste
  • Inclusive on-scale QC ranges provide immediate quality assurance of testing methodology
  • Automated or manual read options are available for ultimate flexibility

Not all products are available for sale in all territories. Please inquire.
 

VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Antimicrobial Agents Various
Antibiotic Code Various
Product Type AST Plate
Target Organism Class Non-fastidious, Gram-positive and Gram-negative Veterinary Isolates
Test Type Veterinary AST, Minimum Inhibitory Concentration (MIC)
Quantity 10/Pk.
Specimen Type Bovine/Porcine Isolates
Species Bovine, Porcine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.