missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sensititre™ ARIS HiQ™ System for AST

Product Code. 18006202
Change view
Click to view available options
Description:
Sensititre ARIS HiQ
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
18006202 Sensititre ARIS HiQ Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18006202 Supplier Thermo Scientific™ Supplier No. YV4000
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Achieve comprehensive, accurate MIC results to guide targeted patient therapy with this benchtop, automated reading and incubation system for AST.

Achieve the comprehensive, accurate MIC values needed to guide more targeted patient therapy with the Thermo Scientific™ Sensititre™ ARIS HiQ™ System, a benchtop automated incubation and reading system for antimicrobial susceptibility testing and identification of bacterial pathogens; designed to accommodate up to 100 Sensititre™ microtitre plates within a single instrument.

Confidently detect emerging antibiotic resistance with the gold-standard level accuracy1,2 of broth microdilution coupled with time-saving benefits of automation to optimize patient care, support antimicrobial stewardship, and improve lab efficiency with the Sensititre™ ARIS HiQ™ System.   

  • Efficient plate handling: central turntable for loading and unloading Sensititre plates via robotic plate transfer and advanced sorting options to optimize read times and efficient plate removal
  • Flexible loading: batch load and unload capability for improved testing workflow
  • Seamless LIS connectivity: Thermo Scientific™ Sensititre SWIN™ software connection is configured to meet LIS connectivity requirements
  • Large capacity, small footprint: to process more tests at one time with limited impact to bench space.

 

The Sensititre ARIS HiQ system helps microbiology laboratories deliver accurate antimicrobial susceptibility testing of the most commonly prescribed antibiotics, as well as novel, last-resort therapies, to provide meaningful results for clinicians, stewardship teams, and public health departments. By providing critical data to support decisions on prescribing the right antibiotic, at the right dose, at the right time, the Sensititre AST System supports the effort to improve patient outcomes and to ensure the right actions are taken to protect the future of healthcare, veterinary, and agriculture industries against the threat of antibiotic resistance.

Not all products are available for sale in all territories. Please inquire.

VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 0°C to 40°C (32°F to 104°F)
Description Sensititre ARIS HiQ
Product Type Microbiology Media
Test Type Minimum Inhibitory Concentration (MIC)
Width (English) 29.53 in.
Width (Metric) 750 mm
Height (English) 44.31 in.
Height (Metric) 1125 mm
Depth (English) 28.94 in.
Depth (Metric) 735 mm
Weight (English) System, empty (no racks, no plates): 90KG (198.5lb)
Weight (Metric) System, full (11 racks - 10 internal racks, 1 utility rack, 100 plates,100μL water per plate): 103KG (226.6lb)
Show More Show Less

Thermo Fisher Scientific products are distributed globally so uses, applications, and availability of product in each country depend on local regulatory marketing authorization status.

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.