missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SENP7 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£385.00 - £587.00
Specifications
| Antigen | SENP7 |
|---|---|
| Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18389986
|
Novus Biologicals
NBP3-17929-25UL |
25 μg |
£385.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18364955
|
Novus Biologicals
NBP3-17929-100UL |
100 μg |
£587.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SENP7 Polyclonal antibody specifically detects SENP7 in Human samples. It is validated for ImmunofluorescenceSpecifications
| SENP7 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| EC 3.4.22.-, KIAA1707SUMO1/sentrin specific protease 7, MGC157730, Sentrin/SUMO-specific protease SENP7, sentrin-specific protease 7, SSP2, SUMO1/sentrin specific peptidase 7, SUMO-1-specific protease 2, SUSP2 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: SERGSQRSKTVDDNSAKQTAHNKEKRRKDDGISLLISDTQPEDLNSGSRGCDHLEQESRNKDVKYSDSKVELTLISRKTKRRLRNNLPDSQYCTSLD | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.2, 40% glycerol | |
| 57337 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title