missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SENP7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582961
This item is not returnable.
View return policy
Description
Immunogen sequence: STISTEFEKP SENYHQDPKL PEEITTKPTK SDFTKLSSLN SQELTLSNAT KSASAGSTTE TVENSNSIDI VGISSLVEKD ENELNTIEKP ILRGHNEGNQ SLISAEPIVV SSDEEGPVEH KSSEILKLQS KQDRE Highest antigen sequence indentity to the following orthologs: Rat - 47%, Mouse - 41%.
SENP7 is a protease that may be involved in two essential functions in the SUMO pathway: processing of full-length SUMO1, SUMO2 and SUMO3 to their mature conjugatable forms and/or deconjugation of SUMO1, SUMO2 and SUMO3 from targeted substrate proteins.
Specifications
| SENP7 | |
| Polyclonal | |
| Unconjugated | |
| SENP7 | |
| 2410152H17Rik; 2810413I22Rik; 2900036C23Rik; 6030449K19Rik; AI790676; KIAA1707; SENP7; sentrin/SUMO-specific protease SENP7; Sentrin-specific protease 7; SSP2; SUMO1/sentrin specific peptidase 7; SUMO1/sentrin specific protease 7; Sumo1/sentrin/SMT3 specific peptidase 7; SUMO-1-specific protease 2; Susp2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57337 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BQF6 | |
| SENP7 | |
| Recombinant protein corresponding to Human SENP7. Recombinant protein control fragment (Product #RP-89269). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction